Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Thermo Scientific Chemicals
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA17878025025 gGlass bottle
Catalog number FSA178780250
Price (MYR)
1,157.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,157.00
Add to cart
Chemical Identifiers
CAS4779-31-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%