Cellulose triacetate
Cellulose triacetate
Cellulose triacetate
Cellulose triacetate
Thermo Scientific Chemicals
Cellulose triacetate
CAS: 9012-09-3 | (C24H32O16)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
Packaging:
Plastic bottle
Catalog number FSA177825000
Price (MYR)
938.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
938.00
Add to cart
Specifications
Chemical Name or MaterialCellulose triacetate
ColorWhite to Yellow
CAS9012-09-3
Physical FormPellets or Rods
TSCATSCA