Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Cellulose acetate, average M.W. 100,000
Thermo Scientific Chemicals
Cellulose acetate, average M.W. 100,000
CAS: 9004-35-7 | (C6H7O5)nC2H3O
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Catalog number FSA177780250
Price (MYR)
377.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
377.00
Add to cart
Chemical Identifiers
CAS504-02-9
IUPAC Namecyclohexane-1,3-dione
Molecular FormulaC6H8O2
InChI KeyHJSLFCCWAKVHIW-UHFFFAOYSA-N
SMILESO=C1CCCC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride content<1.0% (as sodium chloride)
Melting Point100-105?C
FormPowder or chunks
Appearance (Color)White to cream or pale yellow to pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% (dry weight basis)