1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, Thermo Scientific Chemicals
1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, Thermo Scientific Chemicals
1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, Thermo Scientific Chemicals
1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA177485000
Price (MYR)
1,620.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,620.00
Add to cart
Specifications
Chemical Name or Material1, 3-Cyclohexanebis(methylamine)
Name Note99%
CAS2579-20-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger