N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-tert-Butyl-alpha-phenylnitrone, 98%, Thermo Scientific Chemicals
CAS: 3376-24-7 | C11H15NO | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA177350050
Price (MYR)
1,910.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,910.00
Add to cart
Chemical Identifiers
CAS928-92-7
IUPAC Name(4E)-hex-4-en-1-ol
Molecular FormulaC6H12O
InChI KeyVTIODUHBZHNXFP-NSCUHMNNSA-N
SMILESCC=CCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%