Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 L
Packaging:
View all
Plastic Can
Catalog number FSA177150250
Price (MYR)
1,724.00
Add to cart
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Price (MYR)
1,724.00
Add to cart
Chemical Identifiers
CAS108-46-3
IUPAC Namebenzene-1,3-diol
Molecular FormulaC6H6O2
InChI KeyGHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILESOC1=CC(O)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or granules or flakes
Melting point108°C to 112°C
Appearance (Color)White to yellow to beige to pink
Infrared spectrumConforms
HPLC>=97.5 %