D(+)-Sucrose, 99.7%, for biochemistry
D(+)-Sucrose, 99.7%, for biochemistry
D(+)-Sucrose, 99.7%, for biochemistry
D(+)-Sucrose, 99.7%, for biochemistry
Thermo Scientific Chemicals
D(+)-Sucrose, 99.7%, for biochemistry
D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA177142500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Sucrose
CAS57-50-1
Melting Point190.0°C to 192.0°C
ColorColorless
pH6.5 to 7.5 (10% soln.)
View more
General Description
  • D(+)-Sucrose is a disaccharide carbohydrate
  • It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose
Applications
  • D(+)-Sucrose can be used for protein and nucleic acid purification
  • Its fatty acid esters have laboratory research applications in transdermal systems
  • It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
RUO – Research Use Only