Potassium hydrogen phthalate, 99.99%, acidimetric standard, Thermo Scientific Chemicals
Potassium hydrogen phthalate, 99.99%, acidimetric standard, Thermo Scientific Chemicals
Potassium hydrogen phthalate, 99.99%, acidimetric standard, Thermo Scientific Chemicals
Potassium hydrogen phthalate, 99.99%, acidimetric standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydrogen phthalate, 99.99%, acidimetric standard, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA177121000
Price (MYR)
278.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
278.00
Add to cart
Chemical Identifiers
CAS10534-89-1
IUPAC Namecobalt(3+) hexaamine trichloride
Molecular FormulaCl3CoH18N6
InChI KeyJXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILESN.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Appearance (Color)Orange
For ultra trace analysis
RUO – Research Use Only