tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
Packaging:
Glass Bottle
Catalog number FSA177040025
Price (MYR)
460.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
460.00
Add to cart
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Aluminium (Al)=<5 ppm
KMnO4-reducing substance=<0.005 % (as HCOOH)
Appearance (Color)White
Heavy metals (as Pb)=<5 ppm
Phosphate (PO4)=<5 ppm