Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number FSA176840010
Price (MYR)
290.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
290.00
Add to cart
Chemical Identifiers
CAS584-02-1
IUPAC Namepentan-3-ol
Molecular FormulaC5H12O
InChI KeyAQIXEPGDORPWBJ-UHFFFAOYSA-N
SMILESCCC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=97.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4090 to 1.4110 (20°C, 589 nm)
For analysis
RUO – Research Use Only