Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexane, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA176810010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclohexane
Melting Point6.5°C
CAS110-82-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
View more
For analysis
RUO – Research Use Only