Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number FSA176688000
Price (MYR)
2,845.00
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,845.00
Add to cart
Chemical Identifiers
CAS10294-34-5, 75-09-2
Specifications
Chemical Name or MaterialBoron trichloride
ColorColorless to Brown
Name Note1M Solution in Methylene Chloride
CAS10294-34-5
Health Hazard 1Danger
View more
Lewis acid
RUO – Research Use Only