Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number FSA176681000
Price (MYR)
716.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
716.00
Add to cart
Chemical Identifiers
CAS461-72-3
IUPAC Nameimidazolidine-2,4-dione
Molecular FormulaC3H4N2O2
InChI KeyWJRBRSLFGCUECM-UHFFFAOYSA-N
SMILESO=C1CNC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point220°C to 223°C
Titration with NaOH98.5 to 101.5 %
Lewis acid
RUO – Research Use Only