4-(4-Hydroxyphenyl)-2-butanone, 99+%, Thermo Scientific Chemicals
4-(4-Hydroxyphenyl)-2-butanone, 99+%, Thermo Scientific Chemicals
4-(4-Hydroxyphenyl)-2-butanone, 99+%, Thermo Scientific Chemicals
4-(4-Hydroxyphenyl)-2-butanone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(4-Hydroxyphenyl)-2-butanone, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA176472500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9012-09-3
Molecular Formula(C24H32O16)n
SMILESCC(=O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)=O)OC(-*)[C@H](OC(C)=O)[C@H]2OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1O-*
Molecular Weight (g/mol)NaN
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Pellets or rods
Appearance (Color)White to light yellow
Acetyl content43 to 44 wt %