Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals
Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals
Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals
Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals
CAS: 780-69-8 | C12H20O3Si | 240.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA174642500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenyltriethoxysilane
Name Note98%
CAS780-69-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
May cause damage to organs through prolonged or repeated exposure if inhaled.