4-Isopropylaniline, 99%, Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Isopropylaniline, 99%, Thermo Scientific Chemicals
CAS: 99-88-7 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA174530500
Price (MYR)
407.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
407.00
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Appearance (Form)Powder
Loss on drying=<10 %
Molasses number=<390