1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
CAS: 107-08-4 | C3H7I | 169.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA174052500
Price (MYR)
931.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
931.00
Add to cart
Chemical Identifiers
CAS672-15-1
IUPAC Name(2S)-2-amino-4-hydroxybutanoic acid
Molecular FormulaC4H9NO3
InChI KeyUKAUYVFTDYCKQA-VKHMYHEASA-N
SMILESN[C@@H](CCO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Optical Rotation-8.6 ± 1° (c=2 in water)
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%