α-D-Methylglucoside, 99%, Thermo Scientific Chemicals
α-D-Methylglucoside, 99%, Thermo Scientific Chemicals
α-D-Methylglucoside, 99%, Thermo Scientific Chemicals
α-D-Methylglucoside, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
α-D-Methylglucoside, 99%, Thermo Scientific Chemicals
CAS: 97-30-3 | C7H14O6 | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA173841000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.0125mm (0.0005in) thick.
Appearance (Color)Brown
Assay from Supplier's CofA≥99.9% (metals basis)