Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl disulfide, 99%, Thermo Scientific Chemicals
CAS: 882-33-7 | C12H10S2 | 218.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number FSA173710500
Price (MYR)
394.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Chemical Identifiers
CAS66-27-3
IUPAC Namemethyl methanesulfonate
Molecular FormulaC2H6O3S
InChI KeyMBABOKRGFJTBAE-UHFFFAOYSA-N
SMILESCOS(C)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4130 to 1.4150 (20°C, 589 nm)