4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Biphenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA173670100
Price (MYR)
258.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
258.00
Add to cart
Chemical Identifiers
CAS606-45-1
IUPAC Namemethyl 2-methoxybenzoate
Molecular FormulaC9H10O3
InChI KeyPFYHAAAQPNMZHO-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Refractive Index1.5305-1.5345 @ 20?C