ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass bottle
Catalog number FSA173440025
Price (MYR)
2,152.00
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,152.00
Add to cart
Chemical Identifiers
CAS93-85-6
IUPAC Name2-amino-1,3-benzothiazole-6-carboxylic acid
Molecular FormulaC8H6N2O2S
InChI KeyZEAKWWWXCZMODH-UHFFFAOYSA-N
SMILESNC1=NC2=CC=C(C=C2S1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%