ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactone monomer, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA173440010
Price (MYR)
801.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
801.00
Add to cart
Chemical Identifiers
CAS89-86-1
IUPAC Name2,4-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyUIAFKZKHHVMJGS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Cream to light beige to pink
Titration with NaOH>=96.0 %