1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
CAS: 586-78-7 | C6H4BrNO2 | 202.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA173361000
Price (MYR)
1,365.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,365.00
Add to cart
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Metal Impurities≤0.02% (200 ppm)
Assay (special)?2.5 % of Theoretical value (K)
FormPellets