1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
CAS: 586-78-7 | C6H4BrNO2 | 202.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA173360250
Price (MYR)
486.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
486.00
Add to cart
Specifications
Chemical Name or Material1-Bromo-4-nitrobenzene
Name Note99%
CAS586-78-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning