Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D
Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D
Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D
Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D
Thermo Scientific Chemicals
Sodium deuteroxide, for NMR, 30 wt% solution in D2O, 99+ atom % D
CAS: 14014-06-3 | HNaO | 41.003 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA17289020020 gPlastic Bottle
Catalog number FSA172890200
Price (MYR)
644.00
Add to cart
Quantity:
20 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
644.00
Add to cart
Specifications
Chemical Name or MaterialSodium deuteroxide
Density1.4600g/mL
Deuterium (D)99.0% min.
PackagingPlastic Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only