3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-O-Methyldopamine hydrochloride, 99+%, Thermo Scientific Chemicals
CAS: 1477-68-5 | C9H13NO2·HCl | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
Packaging:
Glass bottle
Catalog number FSA172850010
Price (MYR)
1,928.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,928.00
Add to cart
Chemical Identifiers
CAS58-96-8
IUPAC Name1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H12N2O6
InChI KeyDRTQHJPVMGBUCF-UHFFFAOYNA-N
SMILESOCC1OC(C(O)C1O)N1C=CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrophotometry>=98 % (lambda max. 262 nm, pH 7)
Loss on drying=<1 % (105°C, 2 hrs)
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 %
Appearance (Color)White to off-white