Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
CAS: 375-22-4 | C4HF7O2 | 214.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number FSA172801000
Price (MYR)
949.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
949.00
Add to cart
Chemical Identifiers
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment-100+325 Mesh
Appearance (Color)Brown
FormPowder
Total Metal Impurities0.001% max.