Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
CAS: 375-22-4 | C4HF7O2 | 214.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA172800250
Price (MYR)
487.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
487.00
Add to cart
Chemical Identifiers
CAS1838-94-4
IUPAC Name2-ethenyl-2-methyloxirane
Molecular FormulaC5H8O
InChI KeyFVCDMHWSPLRYAB-UHFFFAOYNA-N
SMILESCC1(CO1)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
Refractive Index1.4145-1.4195 @ 20?C