n-Butylboronic acid, 98%, Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA172380050
Price (MYR)
595.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
595.00
Add to cart
Chemical Identifiers
CAS5856-63-3
IUPAC Name2-aminobutan-1-ol
Molecular FormulaC4H11NO
InChI KeyJCBPETKZIGVZRE-UHFFFAOYNA-N
SMILESCCC(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Specific optical rotation-6.5° to -10.5° (20°C, 589 nm) (neat)
GC>=97.5 %
Refractive index1.4495 to 1.4535 (20°C, 589 nm)
View more
Acylation reagent
RUO – Research Use Only