n-Butylboronic acid, 98%, Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA172380010
Price (MYR)
368.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
368.00
Add to cart
Specifications
Chemical Name or Materialn-Butylboronic acid
CAS4426-47-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Acylation reagent
RUO – Research Use Only