Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA172001000
Price (MYR)
311.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
311.00
Add to cart
Chemical Identifiers
CAS3012-65-5
IUPAC Namediammonium 3-carboxy-3-hydroxypentanedioate
Molecular FormulaC6H14N2O7
InChI KeyYXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
Appearance (Form)Crystalline powder or crystals