Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
Packaging:
Plastic bottle
Catalog number FSA172000025
Price (MYR)
2,988.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,988.00
Add to cart
Specifications
Chemical Name or MaterialDimethylglyoxime
Name Note99+%
CAS95-45-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.