p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA171785000
Price (MYR)
249.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
249.00
Add to cart
Chemical Identifiers
CAS87-66-1
IUPAC Namebenzene-1,2,3-triol
Molecular FormulaC6H6O3
InChI KeyWQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILESOC1=CC=CC(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Sulfated ash=<0.1 %
Appearance (Color)White to off-white
HPLC>=98.5 %
Appearance (Form)Crystalline powder