p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 97.5%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA171780010
Price (MYR)
354.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
354.00
Add to cart
Specifications
Chemical Name or Materialp-Toluenesulfonic acid monohydrate
CAS6192-52-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for