Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals
CAS: 701-97-3 | C9H16O2 | 156.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA171450500
Price (MYR)
3,192.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
3,192.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexanepropionic acid
Name Note99%
CAS701-97-3
Health Hazard 1Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation. Harmful if swallowed. Causes skin irritation.