Paraffin, liquid, pure, Thermo Scientific Chemicals
Paraffin, liquid, pure, Thermo Scientific Chemicals
Paraffin, liquid, pure, Thermo Scientific Chemicals
Paraffin, liquid, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraffin, liquid, pure, Thermo Scientific Chemicals
CAS: 8012-95-1 | MFCD00131611
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA171402500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS563-41-7
IUPAC Namehydrogen aminourea chloride
Molecular FormulaCH6ClN3O
InChI KeyXHQYBDSXTDXSHY-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Sulfated ash=<0.1 %
Appearance (Form)Crystalline powder and/or chunks
Titration with KIO3>=99.0 %