meso-Tetraphenylporphine, 97%, Thermo Scientific Chemicals
meso-Tetraphenylporphine, 97%, Thermo Scientific Chemicals
meso-Tetraphenylporphine, 97%, Thermo Scientific Chemicals
meso-Tetraphenylporphine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
meso-Tetraphenylporphine, 97%, Thermo Scientific Chemicals
CAS: 917-23-7 | C44H30N4 | 614.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA171210050
Price (MYR)
1,164.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,164.00
Add to cart
Chemical Identifiers
CAS13818-75-2
IUPAC Namegadolinium(3+) tribromide
Molecular FormulaBr3Gd
InChI KeyKGOKDPWKDBWITQ-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Residual water< 100 ppm
FormPowder
Total Metal Impurities0.01% max.