α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
α-Bromo-p-toluic acid, 97%, Thermo Scientific Chemicals
CAS: 6232-88-8 | C8H7BrO2 | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA170400500
Price (MYR)
1,883.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,883.00
Add to cart
Chemical Identifiers
CAS771-60-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange to brown
FormFused solid
Assay (GC)≥96.0%
Melting Point (clear melt)31.0-40.0?C