Hydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
Hydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
Hydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
Hydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals
Hydroxylamine hydrochloride, 97%, ACROS Organics, Quantity: 1kg, CAS: 5470-11-1, White, Melting Point: 155 deg.C to 158 deg.C, Molecular Formula: H3NO.HCl, Molecular Weight: 69.49, Packaging: Plastic bottle, Percent Purity: 97%
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Catalog number FSA170365000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS124-04-9
IUPAC Namehexanedioic acid
Molecular FormulaC6H10O4
InChI KeyWNLRTRBMVRJNCN-UHFFFAOYSA-N
SMILESOC(=O)CCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point151°C to 153°C
Water=<0.2 %
View more
Reduction reagent
RUO – Research Use Only