alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
Thermo Scientific Chemicals
alpha-D(+)-Glucose, 99+%, anhydrous
CAS: 492-62-6 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA170080010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or Materialα-D(+)-Glucose
Name NoteAnhydrous
Acidity0.15mL 0.1N NaOH/6g max.
CAS492-62-6
CAS Min %99.0