n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA170070025
Price (MYR)
643.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
643.00
Add to cart
Specifications
Chemical Name or Materialn-Pentane
Melting Point-130.0°C
CAS109-66-0
CAS Min %99.0
Health Hazard 1Danger