n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA170070010
Price (MYR)
443.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
443.00
Add to cart
Chemical Identifiers
CAS30433-91-1
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5500 to 1.5540 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Colorless to yellow to orange
Appearance (Form)Liquid