Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA169930025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTributyl phosphate
Name Note99%
CAS126-73-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Suspected of causing cancer.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.