Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 mL
50 mL
Packaging:
Glass bottle
Catalog number FSA169892500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, almost colorless liquid
Specific Absorbance (E 1%/1cm)254nm ≤0.10 AU (measured at 0.1 M concentration)
Specific Absorbance (E 1%/1cm)240nm ≤0.15 AU (measured at 0.1 M concentration)
Specific Absorbance (E 1%/1cm)300nm ≤0.05 AU (measured at 0.1 M concentration)
Assay from Supplier's CofA1.00 ±0.02M