Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals
CAS: 75-11-6 | CH2I2 | 267.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA169835000
Price (MYR)
2,170.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,170.00
Add to cart
Chemical Identifiers
CAS546-89-4
IUPAC Namelithium(1+) acetate
Molecular FormulaC2H3LiO2
InChI KeyXIXADJRWDQXREU-UHFFFAOYSA-M
SMILES[Li+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Appearance (Form)Crystalline powder
Melting point280°C to 285°C