Lauroyl chloride, 98%, Thermo Scientific Chemicals
Lauroyl chloride, 98%, Thermo Scientific Chemicals
Lauroyl chloride, 98%, Thermo Scientific Chemicals
Lauroyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lauroyl chloride, 98%, Thermo Scientific Chemicals
CAS: 112-16-3 | C12H23ClO | 218.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA169032500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLauroyl chloride
CAS112-16-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat