Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA168881000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS871329-52-1
IUPAC Name(2-chloro-3-fluorophenyl)boronic acid
Molecular FormulaC6H5BClFO2
InChI KeyTYOGIUGNZOBBHE-UHFFFAOYSA-N
SMILESOB(O)C1=C(Cl)C(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point113°C to 117°C
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=96.0 %