1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-2-methylpropane, 98+%, Thermo Scientific Chemicals
CAS: 78-77-3 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA168835000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Microbial contamination=<1000 /g (Total plate count)
Microbial contamination=<100 /g (Yeast and Mould)
Microbial contaminationAbsent (E. Coli)
Microbial contaminationAbsent (Salmonella)
Appearance (Color)White to cream to light yellow to beige