Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
CAS: 84-11-7 | C14H8O2 | 208.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA168790250
Price (MYR)
472.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
472.00
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Metallic grey
CommentTitanium: ≥99.5%
FormGauze, 60 mesh woven with 0.009 in. diameter wire