Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals
CAS: 124-41-4 | CH3NaO | 54.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 mL
1 L
10 L
25 L
Catalog number FSA168600025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium methoxide
Name Note30 wt% Solution in Methanol
CAS124-41-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs.
Flammable liquid and vapor.
May be corrosive to metals.